missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PHA 665752, Tocris Bioscience™
Potent and selective MET inhibitor
£305.44 - £1205.16
Chemical Identifiers
CAS | 477575-56-7 |
---|---|
Molecular Formula | C32H34Cl2N4O4S |
Molecular Weight (g/mol) | 641.608 |
InChI Key | OYONTEXKYJZFHA-SSHUPFPWSA-N |
Synonym | unii-0vxu5t5r3j, 2r-1-5-z-5-2,6-dichlorophenyl methyl sulfonyl-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrol-3-yl carbonyl-2-1-pyrrolidinylmethyl pyrrolidine, 0vxu5t5r3j, r,z-5-2,6-dichlorobenzylsulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, pha-665752 hydrate, 3z-5-2,6-dichlorophenyl methanesulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, 3z-5-2,6-dichlorophenyl methylsulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, r,z-5-2,6-dichlorobenzyl sulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, 3z-5-2,6-dichlorophenyl methyl sulfonyl-3-3,5-dimethyl-4-2r-2-1-pyrrolidinylmethyl-1-pyrrolidinyl carbonyl-1h-pyrrol-2-yl methylene-1,3-dihydro-2h-indol-2-one hydrate, d00tbd |
PubChem CID | 10461815 |
IUPAC Name | (3Z)-5-[(2,6-dichlorophenyl)methylsulfonyl]-3-[[3,5-dimethyl-4-[(2R)-2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl]methylidene]-1H-indol-2-one |
SMILES | CC1=C(NC(=C1C(=O)N2CCCC2CN3CCCC3)C)C=C4C5=C(C=CC(=C5)S(=O)(=O)CC6=C(C=CC=C6Cl)Cl)NC4=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18025072
|
Tocris Bioscience
2693/10 |
10 mg |
£305.44
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18095472
|
Tocris Bioscience
2693/50 |
50 mg |
£1205.16
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Potent, selective and ATP-competitive inhibitor of MET kinase (IC50 values are 9, 68, 200, 1400, 3000, 3800 and 6000 nM for MET, Ron, Flk-1, c-abl, FGFR1, EGFR and c-src respectively and > 10000 nM for IGF-IR, PDGFR, AURORA2, PKA, PKBα, p38α, MK2 and MK3). Antitumor agent; inhibits tumorigenicity and angiogenesis in mouse lung cancer xenografts.Chemical Identifiers
477575-56-7 | |
641.608 | |
unii-0vxu5t5r3j, 2r-1-5-z-5-2,6-dichlorophenyl methyl sulfonyl-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrol-3-yl carbonyl-2-1-pyrrolidinylmethyl pyrrolidine, 0vxu5t5r3j, r,z-5-2,6-dichlorobenzylsulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, pha-665752 hydrate, 3z-5-2,6-dichlorophenyl methanesulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, 3z-5-2,6-dichlorophenyl methylsulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, r,z-5-2,6-dichlorobenzyl sulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, 3z-5-2,6-dichlorophenyl methyl sulfonyl-3-3,5-dimethyl-4-2r-2-1-pyrrolidinylmethyl-1-pyrrolidinyl carbonyl-1h-pyrrol-2-yl methylene-1,3-dihydro-2h-indol-2-one hydrate, d00tbd | |
(3Z)-5-[(2,6-dichlorophenyl)methylsulfonyl]-3-[[3,5-dimethyl-4-[(2R)-2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl]methylidene]-1H-indol-2-one |
C32H34Cl2N4O4S | |
OYONTEXKYJZFHA-SSHUPFPWSA-N | |
10461815 | |
CC1=C(NC(=C1C(=O)N2CCCC2CN3CCCC3)C)C=C4C5=C(C=CC(=C5)S(=O)(=O)CC6=C(C=CC=C6Cl)Cl)NC4=O |
Specifications
477575-56-7 | |
unii-0vxu5t5r3j, 2r-1-5-z-5-2,6-dichlorophenyl methyl sulfonyl-1,2-dihydro-2-oxo-3h-indol-3-ylidene methyl-2,4-dimethyl-1h-pyrrol-3-yl carbonyl-2-1-pyrrolidinylmethyl pyrrolidine, 0vxu5t5r3j, r,z-5-2,6-dichlorobenzylsulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, pha-665752 hydrate, 3z-5-2,6-dichlorophenyl methanesulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, 3z-5-2,6-dichlorophenyl methylsulfonyl-3-3,5-dimethyl-4-2r-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylidene-1h-indol-2-one, r,z-5-2,6-dichlorobenzyl sulfonyl-3-3,5-dimethyl-4-2-pyrrolidin-1-ylmethyl pyrrolidine-1-carbonyl-1h-pyrrol-2-yl methylene indolin-2-one, 3z-5-2,6-dichlorophenyl methyl sulfonyl-3-3,5-dimethyl-4-2r-2-1-pyrrolidinylmethyl-1-pyrrolidinyl carbonyl-1h-pyrrol-2-yl methylene-1,3-dihydro-2h-indol-2-one hydrate, d00tbd | |
CC1=C(NC(=C1C(=O)N2CCCC2CN3CCCC3)C)C=C4C5=C(C=CC(=C5)S(=O)(=O)CC6=C(C=CC=C6Cl)Cl)NC4=O | |
641.608 | |
641.61 | |
PHA 665752 |
C32H34Cl2N4O4S | |
OYONTEXKYJZFHA-SSHUPFPWSA-N | |
(3Z)-5-[(2,6-dichlorophenyl)methylsulfonyl]-3-[[3,5-dimethyl-4-[(2R)-2-(pyrrolidin-1-ylmethyl)pyrrolidine-1-carbonyl]-1H-pyrrol-2-yl]methylidene]-1H-indol-2-one | |
10461815 | |
>98% |
Safety and Handling
Recommended Storage : Store at +4°C