missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PHA 665752

Product Code. 18095472
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18095472 50 mg 50mg
18025072 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095472 Supplier Tocris Bioscience™ Supplier No. 2693/50
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective MET inhibitor

Potent, selective and ATP-competitive inhibitor of MET kinase (IC50 values are 9, 68, 200, 1400, 3000, 3800 and 6000 nM for MET, Ron, Flk-1, c-abl, FGFR1, EGFR and c-src respectively and > 10000 nM for IGF-IR, PDGFR, AURORA2, PKA, PKBα, p38α, MK2 and MK3). Antitumor agent; inhibits tumorigenicity and angiogenesis in mouse lung cancer xenografts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 477575-56-7
Molecular Formula C32H34Cl2N4O4S

Specifications

Quantity 50 mg
Formula Weight 641.61
Purity >98%
Molecular Weight (g/mol) 641.608
Chemical Name or Material PHA 665752
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.