missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PHA 568487
GREENER_CHOICE

Product Code. 18054552
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18054552 50 mg 50mg
18015722 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18054552 Supplier Tocris Bioscience™ Supplier No. 3134/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

α7-selective agonist

Agonist of the α7 nicotinic acetylcholine receptor (Ki values are 44 and 2800 for α7 and 5-HT3 respectively, IC50 values are > 100 μM for α3β4 and α1β1δγ respectively, and % inhibition is < 1 and 5% for α4β2 and hERG respectively). Orally active and brain penetrant.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 527680-57-5
Molecular Formula C20H24N2O7

Specifications

Quantity 50 mg
Formula Weight 404.41
Purity >99%
Molecular Weight (g/mol) 404.419
Chemical Name or Material PHA 568487
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.