CAS RN 527680-57-5
CAS RN 527680-57-5
IUPAC Name:
N-{1-azabicyclo[2.2.2]octan-3-yl}-2,3-dihydro-1,4-benzodioxine-6-carboxamide; but-2-enedioic acid
Synonyms:
N-{1-azabicyclo[2.2.2]octan-3-yl}-2,3-dihydro-1,4-benzodioxine-6-carboxamide; butenedioic acid
Molecular Weight (g/mol):
404.42
Molecular Formula:
C20H24N2O7
InChi Key:
QYQZUGYJVHHHEE-UHFFFAOYNA-N
SMILES:
OC(=O)C=CC(O)=O.O=C(NC1CN2CCC1CC2)C1=CC2=C(OCCO2)C=C1
1
–
1
of
1
results
1