Learn More
Cowie Technology™ PTFE Joint Adapter
Cowie Technology™ PTFE Joint Adapters are grease-free and create high vacuum seals for standard taper ground glass joints.
Brand:  Cowie Technology™ 021.1934
Additional Details : Weight : 0.06800kg
 View more versions of this product
Product Code. 10271852
  • £18.75 / Each
Estimated Shipment: 02-07-2024
to see stock.
Add to basket
Description
Description
  • Grease-free
  • High vacuum seals for standard taper ground glass joints
  • Substantial shoulder for ease of handling and repeated use
  • High vacuum performance (leakage rate is less than 1 x 10-4 toor, litre, sec.-1) is attained using transverse sealing rings.
  • Use up to 280°C
Specifications
Specifications
1 Each
34/35
NS 19/26, NS 34/35
PTFE Joint Adapters
19/26