missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Specifications
| Certifications/Compliance | ISO 9001 |
| pH Range | 5.5 to 9 |
| Format | Roll |
| Graduations | 5.5, 6.0, 6.5, 7.0, 7.5, 8.0, 8.5, 9.0 |
| Quantity | 1 Each |
| Type | pH Paper |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?