missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ pH Paper
Shop All Macherey-Nagel Products
missing translation for 'orderingAttributeHoverText'
pH Range:
0.5 to 5.5
1 to 11
1 to 14
3.8 to 5.8
4 to 7
5.4 to 7
5.5 to 9
6.4 to 8
7.2 to 9.7
9 to 13
missing translation for 'unitSize'
5m
Each
Pack of 5
Description
Measure pH simply and easily. Macherey-Nagel™ pH paper comes in a circular format with a wide variety of pH ranges.
- Universal and special indicator papers - simple pH measurement
- Universal and special indicator papers supplied in plastic reels that ensure protection against external influences .
- Always ready for use .
- The reaction color of the paper is compared with the color scale and read the pH . Allows a very simple pH measurement .
Specifications
Specifications
| Certifications/Compliance | ISO 9001 |
| pH Range | 3.8 to 5.8 |
| Format | Roll |
| Graduations | <3.8, 3.8, 4.1, 4.3, 4.5, 4.7, 4.9, 5.2, 5.5, 5.8 >5.8 |
| Quantity | 1 Each |
| Type | pH Paper |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction