missing translation for 'onlineSavingsMsg'
Learn More

Hanna Instruments™ pH/ORP/ISE/EC/TDS/NaCl Benchtop Meter

Product Code. 12923267
Change view
Click to view available options
Dimensions (L x W x H):
235 x 218 x 108mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W x H)
12923267 235 x 218 x 108mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12923267 Supplier Hanna Instruments™ Supplier No. HI2550

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Accuracy (ISE) ±0.2/±1mV
Accuracy (mV) ±0.2/±1mV
Accuracy (pH) ±0.01/±0.002
Accuracy (TDS) ±1% of Reading
Calibration Points Up to five point calibration, seven standard buffers available and two custom buffers
Calibration Points (pH) Up to five point calibration, seven standard buffers available and two custom buffers
Detection Range –2 to 16.00°C
Channels 2
Display Type LCD
Range (mV) ±999.9 mV, ±2000 mV
Range (pH) -2 to 16.00
Resolution (ISE) 0.1/1 mV
Resolution (pH) 0.1/0.01/0.001
Temperature Range (Metric) -20°C to +120.0°C
Inputs BNC
Model HI-2550
Power Supply 12 VDC
Product Type pH/ORP/ISE/EC/TDS/NaCl Benchtop Meter
Range (ISE) ±999.9/±2000mV
Range (ORP) ±999.9/±2000mV
Resolution (mV) 0.1/1mV
Resolution (ORP) 0.1/1mV
TDS Factor Range 0.8
Temperature Accuracy ±0.4°C
Temperature Compensation –20 to 120.0°C
Temperature (Metric) Operating -20°C to +120.0°C
Temperature Probe Calibration HI 7662 temperature probe with 1 m (3.3') cable (included)
Dimensions (L x W x H) 235 x 218 x 108mm
Voltage 230 V
Frequency 50Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.