missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ pH Electrode

Product Code. 11759748 Shop All Fisherbrand Traceable Products
Change view
Click to view available options
Length (Metric):
102 mm
115 mm
120 mm
152 mm
225 mm
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11759748 102 mm Each
11785628 115 mm Each
11755618 120 mm Each
11765618 225 mm Each
11739738 152 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 11759748 Supplier Fisherbrand™ Supplier No. 1932527010B
  • ONLINE EXCLUSIVE PRICE
    £18.68
    £9.65 / Pack of 1
    Save £9.03
    48% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

This unique maintenance free solid electrolyte pH electrode for measurement up to 110 ° C and 10 bar, is ideal for industrial applications.

  • Maintenance-free pH electrode with 1/2 "MNPT thread for mounting
  • Max. 6 bar
  • Electrode for measuring special pool water
  • Pt1000 temperature sensor integrated 
  • Supplied with 3 m cable and BNC plug

Specifications

Length (Metric) 102 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.