missing translation for 'onlineSavingsMsg'
Learn More

PGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

£135.00 - £349.00

Specifications

Antigen PGM1
Concentration 0.39 mg/mL
Applications Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16814224
View Documents
Proteintech
15161-1-AP-20UL
20 μL
£135.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
16804224
View Documents
Proteintech
15161-1-AP-150UL
150 μL
£349.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Phosphoglucomutases.
Specifications

Specifications

PGM1
Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Rabbit
Human, Rat, Mouse
P36871, P38652, Q9D0F9
24645, 5236, 66681
PGM1 Fusion Protein Ag7269
Primary
-20°C
PGM1
0.39 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1
PGM1
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.