missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGM1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15161-1-AP-20UL
This item is not returnable.
View return policy
Description
Phosphoglucomutases.Specifications
| PGM1 | |
| Polyclonal | |
| Unconjugated | |
| PGM1 | |
| Glucose phosphomutase 1, PGM 1, PGM1, phosphoglucomutase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 24645, 5236, 66681 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Flow Cytometry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P36871, P38652, Q9D0F9 | |
| PGM1 | |
| PGM1 Fusion Protein Ag7269 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction