missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGD Mouse anti-Human, Unconjugated, Clone: 2A10G4, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PGD |
|---|---|
| Clone | 2A10G4 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17462413
|
Proteintech
67916-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17452413
|
Proteintech
67916-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The PGD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PGD. This antibody recognizes human antigen. The PGD antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PGD | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Human | |
| IgG1 |
| 2A10G4 | |
| Monoclonal | |
| Mouse | |
| 5226 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title