Learn More
PGD Mouse anti-Human, Unconjugated, Clone: 2A10G4, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67916-1-IG-150UL
This item is not returnable.
View return policy
Description
The PGD antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PGD. This antibody recognizes human antigen. The PGD antibody has been validated for the following applications: IF, WB, ELISA analysis.
Specifications
| PGD | |
| Monoclonal | |
| Unconjugated | |
| 150 μL | |
| 5226 | |
| IgG1 |
| Western Blot, Immunofluorescence | |
| 2A10G4 | |
| Mouse | |
| Primary | |
| Human |