missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM5 Mouse anti-Human, Mouse, Pig, Rabbit, Rat, Clone: 1C8B9, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | PGAM5 |
|---|---|
| Clone | 1C8B9 |
| Concentration | 1000 μg/mL |
| Applications | Immunohistochemistry, Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17761169
|
Proteintech
68116-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17751169
|
Proteintech
68116-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGAM5 Monoclonal antibody specifically detects PGAM5 in Human, Mouse, Pig, Rabbit, Rat samples. It is validated for IHC, WB, ELISASpecifications
| PGAM5 | |
| 1000 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Pig, Rabbit, Rat | |
| 192111 | |
| Primary | |
| Store at -20°C |
| 1C8B9 | |
| Immunohistochemistry, Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| IgG1 | |
| Protein G purification | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title