missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PGAM5 Mouse anti-Human, Mouse, Pig, Rabbit, Rat, Clone: 1C8B9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68116-1-IG-20UL
This item is not returnable.
View return policy
Description
PGAM5 Monoclonal antibody specifically detects PGAM5 in Human, Mouse, Pig, Rabbit, Rat samples. It is validated for IHC, WB, ELISASpecifications
| PGAM5 | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
| Mouse | |
| 20 μL | |
| 192111 | |
| Store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, ELISA | |
| 1C8B9 | |
| Unconjugated | |
| BC136328 | |
| Protein G purification | |
| Primary | |
| Human, Mouse, Pig, Rabbit, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction