missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGAM1 Recombinant Rabbit Monoclonal Antibody (24GB1635)

Rabbit Recombinant Monoclonal Antibody
£172.00 - £488.00
Specifications
| Antigen | PGAM1 |
|---|---|
| Clone | 24GB1635 |
| Content And Storage | -20°C |
| Applications | Flow Cytometry, Western Blot |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30275225
![]() |
Invitrogen™
MA557574 |
100 μL |
£488.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30275437
|
Invitrogen™
MA557575 |
20 μL |
£172.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PGAM1 Recombinant Monoclonal Antibody for Western Blot
PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombinant human PGAM protein, fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography techniques.Specifications
| PGAM1 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| PGAM1 | |
| 2310050F24Rik; BPG-dependent PGAM 1; CDABP0006; epididymis secretory protein Li 35; HEL-S-35; PGAM1; Pgam-1; PGAMA; PGAM-B; Pgmut; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase isozyme B; RP11-452K12.8; similar to phosphoglycerate mutase | |
| PGAM1 | |
| Primary | |
| Affinity chromatography |
| 24GB1635 | |
| Flow Cytometry, Western Blot | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18669, P25113, Q9DBJ1 | |
| 18648, 24642, 5223 | |
| A synthesized peptide derived from human PGAM1 (aa 1-54). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title