missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PGAM1 Recombinant Rabbit Monoclonal Antibody (24GB1635)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557575
This item is not returnable.
View return policy
Description
PGAM1 Recombinant Monoclonal Antibody for Western Blot
PGAM1 belongs to the phosphoglycerate mutase family. PGAM1 is an important component of glucose and 2,3-BPGA (2,3-bisphosphoglycerate) metabolism and catalyzes the reversible reaction of 3-phosphoglycerate (3-PGA) to 2-phosphoglycerate (2-PGA) in the glycolytic pathway. The PGAM1 protein is a dimeric enzyme containing, in different tissues, different proportions of a slow-migrating muscle (MM) isozyme, a fast-migrating brain (BB) isozyme, and a hybrid form (MB). Mutations in PGAM1 cause muscle phosphoglycerate mutase efficiency, also known as glycogen storage disease X. Recombinant human PGAM protein, fused to His-tag at N-terminus, was expressed in E. coli and purified by using conventional chromatography techniques.
Specifications
| PGAM1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| PGAM1 | |
| 2310050F24Rik; BPG-dependent PGAM 1; CDABP0006; epididymis secretory protein Li 35; HEL-S-35; PGAM1; Pgam-1; PGAMA; PGAM-B; Pgmut; Phosphoglycerate mutase 1; phosphoglycerate mutase 1 (brain); phosphoglycerate mutase A, nonmuscle form; phosphoglycerate mutase isozyme B; RP11-452K12.8; similar to phosphoglycerate mutase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 18648, 24642, 5223 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot | |
| 24GB1635 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P18669, P25113, Q9DBJ1 | |
| PGAM1 | |
| A synthesized peptide derived from human PGAM1 (aa 1-54). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction