missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PG 01037 dihydrochloride
GREENER_CHOICE

Product Code. 18075722
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18075722

Brand: Tocris Bioscience™ 3887/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

D3 receptor selective antagonist

Dopamine D3 receptor antagonist; 133-fold selective for D3 over D2 receptors in vitro (Ki values are 0.70, 93.3 and 375 nM for D3, D2 and D4 receptors respectively). Attenuates abnormal involuntary movements associated with L-DOPA (Cat. No. 3788) in rat models of Parkinson's disease. Inhibits the effects of methamphetamine; attenuates drug-induced behaviors in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 675599-62-9
Molecular Formula C26H28Cl4N4O

Specifications

Quantity 10 mg
Formula Weight 554.34
Purity >99%
Molecular Weight (g/mol) 554.337
Chemical Name or Material PG 01037 dihydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.