accessibility menu, dialog, popup

UserName

CAS RN 675599-62-9

NNNHONClClHClHCl(E/Z)

CAS RN 675599-62-9

IUPAC Name: N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]but-2-en-1-yl}-4-(pyridin-2-yl)benzamide dihydrochloride
Synonyms: N-{4-[4-(2,3-dichlorophenyl)piperazin-1-yl]but-2-en-1-yl}-4-(pyridin-2-yl)benzamide dihydrochloride
Molecular Weight (g/mol): 554.34
Molecular Formula: C26H28Cl4N4O
InChi Key: DACFFMAXBWGOKQ-UHFFFAOYSA-N
SMILES: Cl.Cl.ClC1=CC=CC(N2CCN(CC=CCNC(=O)C3=CC=C(C=C3)C3=CC=CC=N3)CC2)=C1Cl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results