missing translation for 'onlineSavingsMsg'
Learn More

Bohlender™ BOLA™ PFA Flexible Tubing

Use this tubing with BOLA Laboratory Screw Joints for a tight, vacuum suitable connection. Bohlender™ BOLA™ PFA Flexible Tubing features circular corrugations around the longitudinal axis and cylindrical tubing ends which can connect directly to fittings, stopcocks or hose connectors.

Brand:  Bohlender™ S1822-28

 View more versions of this product

Product Code. 10660942

  • £297.86 / Each
Estimated Shipment: 22-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flexible to highly flexible corrugated tubing
  • Tight bending radius causes little cross-section reduction
  • Non-porous
  • Translucent
Specifications

Specifications

FDA Certified
PFA
1 m
For connections under vibrations
1 Pack
Flexible
260°C
14 mm
16 mm
1 mm
-270°C
Transparent
2 bar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers