missing translation for 'onlineSavingsMsg'
Learn More

PF 477736, Tocris Bioscience™
GREENER_CHOICE

Product Code. 18035302
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18035302 10 mg 10mg
18035622 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18035302 Supplier Tocris Bioscience Supplier No. 4277/10

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Selective Chk1 inhibitor

Selective checkpoint kinase 1 (Chk1) inhibitor (Ki values are 0.49 and 47 nM for Chk1 and Chk2 respectively). Abrogates cell cycle arrest at S and G2-M checkpoints; sensitizes cells to DNA damage. Enhances docetaxel (Cat. No. 4056) efficacy in tumor cells and xenografts.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 952021-60-2
Molecular Formula C22H25N7O2
Molecular Weight (g/mol) 419.489
InChI Key YFNWWNRZJGMDBR-LJQANCHMSA-N
Synonym unii-xo23pgz0sm, xo23pgz0sm, 2r-2-amino-2-cyclohexyl-n-2-1-methyl-1h-pyrazol-4-yl-6-oxo-5,6-dihydro-1h-1,2 diazepino 4,5,6-cd indol-8-yl-acetamide, r-2-amino-2-cyclohexyl-n-2-1-methyl-1h-pyrazol-4-yl-6-oxo-5,6-dihydro-1h-1,2 diazepino 4,5,6-cd indol-8-yl acetamide, 2r-2-amino-2-cyclohexyl-n-2-1-methylpyrazol-4-yl-9-oxo-3,10,11-triazatricyclo 6.4.1.0?, 1 3 trideca-1,4,6,8 13 ,11-pentaen-6-yl acetamide, d0u9iq, cyclohexaneacetamide, alpha-amino-n-5,6-dihydro-2-1-methyl-1h-pyrazol-4-yl-6-oxo-1h-pyrrolo 4,3,2-ef 2,3 benzodiazepin-8-yl-, alphar, pf hplc, ar-, a-amino-n-5,6-dihydro-2-1-methyl-1h-pyrazol-4-yl-6-oxo-1h-pyrrolo 4,3,2-ef 2,3 benzodiazepin-8-yl cyclohexaneacetamide, 2∼ r-2-azanyl-2-cyclohexyl-∼ n-2-1-methylpyrazol-4-yl-9-oxidanylidene-3,10,11-triazatricyclo 6.4.1.0^ 4,13 trideca-1,4,6,8 13 ,11-pentaen-6-yl ethanamide
PubChem CID 16750408
SMILES CN1C=C(C=N1)C2=NC3=C4C2=CNNC(=O)C4=CC(=C3)NC(=O)C(C5CCCCC5)N

Specifications

Quantity 10 mg
Formula Weight 419.48
Percent Purity >99%
Chemical Name or Material PF 477736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.