accessibility menu, dialog, popup

UserName

CH3NNNNHNHOHNOH2N

CAS RN 952021-60-2

IUPAC Name: 2-amino-2-cyclohexyl-N-[2-(1-methyl-1H-pyrazol-4-yl)-9-oxo-3,10,11-triazatricyclo[6.4.1.0⁴,¹³]trideca-1(12),2,4,6,8(13)-pentaen-6-yl]acetamide
Synonyms: 2-amino-2-cyclohexyl-N-[2-(1-methylpyrazol-4-yl)-9-oxo-3,10,11-triazatricyclo[6.4.1.0⁴,¹³]trideca-1(12),2,4,6,8(13)-pentaen-6-yl]acetamide
Molecular Weight (g/mol): 419.49
Molecular Formula: C22H25N7O2
InChi Key: YFNWWNRZJGMDBR-UHFFFAOYNA-N
SMILES: CN1C=C(C=N1)C1=NC2=CC(NC(=O)C(N)C3CCCCC3)=CC3=C2C1=CNNC3=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results