missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ PF 3845
GREENER_CHOICE

Product Code. 18063872
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18063872 50 mg 50mg
18005002 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18063872 Supplier Tocris Bioscience™ Supplier No. 4175/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective FAAH inhibitor

Selective fatty acid amide hydrolase (FAAH) inhibitor (Ki = 0.23 μM). Reduces inflammatory pain via a cannabinoid receptor-dependent mechanism. Highly efficacious and selective in vivo. Displays no activity at FAAH-2 (IC50 >10 μM).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1196109-52-0
Molecular Formula C24H23F3N4O2

Specifications

Quantity 50 mg
Formula Weight 456.46
Purity >99%
Molecular Weight (g/mol) 456.47
Chemical Name or Material PF 3845
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.