missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PEX5L Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91484
This item is not returnable.
View return policy
Description
PEX5L Polyclonal specifically detects PEX5L in Mouse samples. It is validated for Western Blot.
Specifications
| PEX5L | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| PEX5-related protein, peroxisomal biogenesis factor 5-like, PEX5R, PEX5RP, PXR2, PXR2B | |
| Rabbit | |
| 68 kDa | |
| 100 μL | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| NP_067458 | |
| PEX5L | |
| Synthetic peptide directed towards the C terminal of human Pex2. Peptide sequence QRKSRNQQQVPHPAISGNIWAALRIALSLMDQPELFQAANLGDLDVLLRA. | |
| Affinity purified | |
| RUO | |
| 51555 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction