Learn More
Agilent BioTek Petri Dish Adapter
Product Code. p-7235923 Shop All Agilent BioTek Products
Click to view available options
Holds:
2 Petri Dishes
4 Dishes
4 Petri Dishes
Unit Size:
Each
Product Code. 16532182
Brand: Agilent BioTek 02748
Petri dish adapters are for use with various size petri dishes and Microplate Washer and Reader Instruments.
  • All positions compatible with imaging up to 20x magnification
  • Position 1 compatible with 40x and 60x magnification
TRUSTED_SUSTAINABILITY
Specifications
Product Type Petri Dish Adapter
For Use With (Equipment) BD Falcon 35 mm (PN: 351008) or Similar Petri Dish up to 38.7 mm maximum diameter; Agilent BioTek Microplate Washer and Reader Instruments
Diameter (Metric) Each Position: 38.7 mm (max.)
Holds 4 Petri Dishes