Learn More
Agilent BioTek Petri Dish Adapter
Shop All Agilent BioTek Products
Click to view available options
Holds:
2 Petri Dishes
4 Dishes
4 Petri Dishes
Unit Size:
Each
Description
- All positions compatible with imaging up to 20x magnification
- Position 1 compatible with 40x and 60x magnification
Specifications
Specifications
| Product Type | Petri Dish Adapter |
| For Use With (Equipment) | BD Falcon 35 mm (PN: 351008) or Similar Petri Dish up to 38.7 mm maximum diameter; Agilent BioTek Microplate Washer and Reader Instruments |
| Diameter (Metric) | Each Position: 38.7 mm (max.) |
| Holds | 4 Petri Dishes |