missing translation for 'onlineSavingsMsg'
Learn More
Learn More
peroxiredoxin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | peroxiredoxin 2 |
|---|---|
| Concentration | 0.2 mg/mL |
| Applications | Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16896993
|
Proteintech
10545-2-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16886993
|
Proteintech
10545-2-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8 (+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2. 7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene.Specifications
| peroxiredoxin 2 | |
| Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human, Rat | |
| P32119, P35704, Q61171 | |
| 21672, 29338, 7001 | |
| peroxiredoxin 2 Fusion Protein Ag0835 | |
| Primary | |
| -20°C | |
| PRDX2 |
| 0.2 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| NKEF B, NKEFB, peroxiredoxin 2, PRDX2, PRP, PRX II, PRX2, PRXII, TDPX1, Thioredoxin peroxidase 1, TSA | |
| PRDX2 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title