Learn More
peroxiredoxin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10545-2-AP-20UL
 View more versions of this product
Product Code. 16896993
  • £135.00 / 20µL
Description
This gene encodes a member of the peroxiredoxin family of antioxidant enzymes, which reduce hydrogen peroxide and alkyl hydroperoxides. The encoded protein may play an antioxidant protective role in cells, and may contribute to the antiviral activity of CD8 (+) T-cells. This protein may have a proliferative effect and play a role in cancer development or progression. The crystal structure of this protein has been resolved to 2. 7 angstroms. Transcript variants encoding distinct isoforms have been identified for this gene.
Specifications
Specifications
peroxiredoxin 2
Polyclonal
Unconjugated
PRDX2
NKEF B, NKEFB, peroxiredoxin 2, PRDX2, PRP, PRX II, PRX2, PRXII, TDPX1, Thioredoxin peroxidase 1, TSA
Rabbit
Antigen Affinity Chromatography
RUO
21672, 29338, 7001
-20°C
Liquid
Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P32119, P35704, Q61171
PRDX2
peroxiredoxin 2 Fusion Protein Ag0835
20 μL
Primary
Mouse, Human, Rat