Learn More
Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips
Volume ranges from 20 to 200μL
Brand:  Axygen™ RRP-20-C-R
Code : 68
Additional Details : Weight : 11.30000kg
 View more versions of this product
Product Code. 12752916
  • £820.00 / Pack of 4800
Estimated Shipment: 10-09-2024
to see stock.
Add to basket
Description
Description
  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts
Order Info
96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case
Specifications
Specifications
20 μL
Non-sterile
Blue
DNase, RNase, and Pyrogen free
Automation, Round
50 Racks/Cs.
Polypropylene
Non-Filtered
Clear
Autoclavable
Rack
SBS Rack
PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head