missing translation for 'onlineSavingsMsg'
Learn More

Axygen™ PerkinElmer™ Janus™ & Multiprobe Robotic Tips

Product Code. 12585477 Shop All Axygen Scientific Products
Click to view available options
Volume (Metric):
20 μL
25 μL
50 μL
175 μL
200 μL
Sterility:
Non-sterile
Sterile
Color:
Black
Clear
Unit Size:
Pack of 4800
Pack of 960
This item is not returnable. View return policy

Product Code. 12585477

Brand: Axygen™ RRP20CRS

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Volume ranges from 20 to 200μL

  • Racked
  • Black and Clear tips available
  • With Liquid level sensing
  • Nonsterile and Presterilized tips
  • Blue, Red and Green rack color inserts

Order Info

96 Tips/Rack, 10 Racks/Pack, 5 Packs/Case

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 20 μL
Sterility Sterile
Color Clear
Autoclavable Autoclavable
Clean Claims DNase, RNase, and Pyrogen free
Format Rack
Tip Style Automation, Round
For Use With (Equipment) PerkinElmer Janus, PerkinElmer MultiProbe Variospan-1, 4, 8 Head
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.