missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripheral Node Addressin Antibody (MECA-79) - Low Endotoxin and Azide Free, Novus Biologicals™
Rat Monoclonal Antibody
£210.00 - £440.00
Specifications
| Antigen | Peripheral Node Addressin |
|---|---|
| Clone | MECA-79 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476) |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266716
|
Novus Biologicals
NBP2-78790SS |
0.1 mg |
£210.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18269927
|
Novus Biologicals
NBP2-78790 |
0.5 mg |
£440.00
0.50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.Specifications
| Peripheral Node Addressin | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| MECA-79, Peripheral Node Addressin, PNAd | |
| IgM κ | |
| IgM purified |
| MECA-79 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476) | |
| Monoclonal | |
| Purified | |
| Lipid and Metabolism | |
| PBS with No Preservative | |
| Mouse lymph node stromal cells | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title