missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peripheral Node Addressin Antibody (MECA-79) - Low Endotoxin and Azide Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-78790
This item is not returnable.
View return policy
Description
Peripheral Node Addressin Monoclonal specifically detects Peripheral Node Addressin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vivo assay.
Specifications
| Peripheral Node Addressin | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:100-1:500, Immunohistochemistry-Frozen 1:100-1:500, In vivo assay reported in scientific literature (PMID 30277476) | |
| MECA-79, Peripheral Node Addressin, PNAd | |
| Mouse lymph node stromal cells | |
| 0.5 mg | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgM κ |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| MECA-79 | |
| Unconjugated | |
| PBS with No Preservative | |
| Rat | |
| IgM purified | |
| Lipid and Metabolism | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction