missing translation for 'onlineSavingsMsg'
Learn More

Perfluoro-1,2-dimethylcyclobutane, 97%, remainder 1,3-isomer, Thermo Scientific Chemicals

Product Code. 11319385
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11319385 25 g 25g
11329385 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 11319385 Supplier Thermo Scientific Alfa Aesar Supplier No. L16757.14
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Perfluoro-1,2-dimethylcyclobutane acts as a ligand for Suzuki coupling. It is involved in the infrared multiple-photon decomposition study of its decomposition products induced by the transverse excited atmospheric pressure pulsed carbon dioxide laser (TEA CO2laser) irradiation.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2994-71-0
Molecular Formula C6F12
Molecular Weight (g/mol) 300.05
MDL Number MFCD00013737
InChI Key RBTROQHBNLSUTL-UHFFFAOYNA-N
Synonym Dodecafluorodimethylcyclobutane
PubChem CID 12789874
SMILES FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F

Specifications

Melting Point -32°C
Density 1.62
Boiling Point 45°C
Refractive Index 1.254
Quantity 25 g
Solubility Information Immiscible with water.
IUPAC Name (3R,4R)-1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Formula Weight 300.05
Percent Purity 97%
Assay remainder 1,3-isomer
Chemical Name or Material Perfluoro-1,2-dimethylcyclobutane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.