accessibility menu, dialog, popup

UserName

FFFFFFFFFFFF

CAS RN 2994-71-0

IUPAC Name: 1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Synonyms: 1,1,2,2,3,4-hexafluoro-3,4-bis(trifluoromethyl)cyclobutane
Molecular Weight (g/mol): 300.05
Molecular Formula: C6F12
InChi Key: RBTROQHBNLSUTL-UHFFFAOYNA-N
SMILES: FC(F)(F)C1(F)C(F)(F)C(F)(F)C1(F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results