missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Staphylococcus aureus V-8 Protease
Shop All Thermo Scientific Products
Click to view available options
Quantity:
5 mg
Unit Size:
Each
Description
Thermo Scientific™ Pierce V-8 Protease Kit contains Staphylococcus aureus V-8 protease immobilized on resin and three digestion buffers for specific cleavage at glutamic acid residues or cleavage at glutamic and aspartic acid residues.
Has two pH optima, one at pH 4.0 and the other at pH 7.8 and is specific for cleavage at the carboxy terminus of glutamic acid and aspartic acid residues. The protease is further specific for only glutamic digestion in buffers, which do not contain phosphate at either pH optimum. Phosphate buffers at pH 7.8 will facilitate digestion at both glutamic and aspartic acid residues.
Highlights:
- Specificity for glutamic acid is achieved in ammonium bicarbonate, pH 7.8 or ammonium acetate, pH 4
- Enzyme is active in the presence of many denaturing agents such as SDS, urea and guanidine·HCl
Specifications
Specifications
| Description | Pierce Staphylococcus aureus V-8 Protease |
| Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
| Form | Solid |
| Quantity | 5 mg |
| Product Type | Protease |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction