missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Staphylococcus aureus V-8 Protease and Kt

Product Code. 10539083 Shop All Thermo Scientific Products
Click to view available options
Quantity:
2.5 mL Resin Kit
Unit Size:
1 set
This item is not returnable. View return policy

Product Code. 10539083

Brand: Thermo Scientific™ 20151

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifically cleave only at glutamic acid residues or cleave at both glutamic and aspartic residues with our immobilized Staphylococcus aureus V-8 protease kit.

Thermo Scientific™ Pierce V-8 Protease Kit contains Staphylococcus aureus V-8 protease immobilized on resin and three digestion buffers for specific cleavage at glutamic acid residues or cleavage at glutamic and aspartic acid residues.

Has two pH optima, one at pH 4.0 and the other at pH 7.8 and is specific for cleavage at the carboxy terminus of glutamic acid and aspartic acid residues. The protease is further specific for only glutamic digestion in buffers, which do not contain phosphate at either pH optimum. Phosphate buffers at pH 7.8 will facilitate digestion at both glutamic and aspartic acid residues.

Highlights:

  • Specificity for glutamic acid is achieved in ammonium bicarbonate, pH 7.8 or ammonium acetate, pH 4
  • Enzyme is active in the presence of many denaturing agents such as SDS, urea and guanidine·HCl
TRUSTED_SUSTAINABILITY

Specifications

Description Pierce Staphylococcus aureus V-8 Protease Kit
Formulation

Multi-component kit with agarose resin (activity 35-60 units/mL), buffers and accessories

Kit Contents V-8 Digestion Buffer 3, 200mL,

Resin Separators, 6

, V-8 Digestion Buffer 2, 200mL, Immobilized S. aureus Protease, 2.5mL, V-8 Digestion Buffer 1, 200mL
MS-Compatible No
Sufficient For 20 digestions of 2mg protein samples
Content And Storage 4°C
Quantity 2.5 mL Resin Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.