missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptide YY Antibody (4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£156.00 - £336.00
Specifications
| Antigen | Peptide YY |
|---|---|
| Clone | 4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, ELISA 1:100-1:2000 |
| Applications | ELISA, Western Blot |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18161534
|
Novus Biologicals
NBP2-23584-0.025ML |
0.025 mL |
£156.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18162072
|
Novus Biologicals
NBP2-23584-0.1ML |
0.1 mL |
£336.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA.Specifications
| Peptide YY | |
| 1 mg/mL | |
| ELISA, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
| PYY | |
| IgG1 κ | |
| Protein A or G purified | |
| Clone 4 binds human PYY(3-36) Based on inhibition ELISA, Clone 4 shows no cross-reaction with PYY(1-36), PYY from mouse, rat, dog or pig, or NPY. |
| 4 | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| 5697 | |
| Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier | |
| Primary | |
| Store at 4C. Do not freeze. |
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel