missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Peptide YY Antibody (4) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-23584-0.1ML
This item is not returnable.
View return policy
Description
Peptide YY Monoclonal specifically detects Peptide YY in Human samples. It is validated for Western Blot, ELISA.
Specifications
| Peptide YY | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Peptide tyrosine tyrosine, peptide YY, PYY1, PYY-I | |
| Mouse | |
| Protein A or G purified | |
| Signal Transduction | |
| 5697 | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| 4 | |
| Unconjugated | |
| 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide | |
| PYY | |
| Synthetic N-terminal fragment of human PYY(3-36) coupled to carrier | |
| 0.1 mL | |
| Primary | |
| Clone 4 binds human PYY(3-36) Based on inhibition ELISA, Clone 4 shows no cross-reaction with PYY(1-36), PYY from mouse, rat, dog or pig, or NPY. | |
| Store at 4C. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction