missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen C/PGC/Progastricsin Antibody (90/11), Novus Biologicals™
Mouse Monoclonal Antibody
£170.00 - £378.00
Specifications
| Antigen | Pepsinogen C/PGC/Progastricsin |
|---|---|
| Clone | 90/11 |
| Concentration | 1.0 mg/mL |
| Dilution | ELISA 1:100-1:2000 |
| Applications | ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18081876
|
Novus Biologicals
NB100-62819-0.025MG |
0.025 mg |
£170.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18066264
|
Novus Biologicals
NB100-62819 |
0.1 mg |
£378.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.Specifications
| Pepsinogen C/PGC/Progastricsin | |
| 1.0 mg/mL | |
| ELISA | |
| Unconjugated | |
| RUO | |
| EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C) | |
| PGA3 | |
| IgG1 | |
| Affinity Purified | |
| NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol |
| 90/11 | |
| ELISA 1:100-1:2000 | |
| Monoclonal | |
| Mouse | |
| Human | |
| 643834 | |
| Human pepsinogen II | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title