missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen C/PGC/Progastricsin Antibody (90/11), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62819-0.025MG
This item is not returnable.
View return policy
Description
Pepsinogen C/PGC/Progastricsin Monoclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.
Specifications
| Pepsinogen C/PGC/Progastricsin | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1:100-1:2000 | |
| EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C) | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 643834 | |
| Human | |
| IgG1 |
| ELISA | |
| 90/11 | |
| Unconjugated | |
| Phosphate/Citrate Buffered Saline with 0.09% Sodium Azide | |
| PGA3 | |
| Human pepsinogen II | |
| 0.025 mg | |
| Primary | |
| NB100-62819 is specific for human pepsinogen II. Less than 1% reactivity has been observed with human pepsinogen I (isozymogens 2-5), porcine pepsinogen, gastrin I, somatostatin, peptide YY and bombesin. Affinity: 1 x 109 l/mol | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction