missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEPD Recombinant Rabbit Monoclonal Antibody (23GB2830), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£179.00 - £485.00
Specifications
| Antigen | PEPD |
|---|---|
| Clone | 23GB2830 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089737
![]() |
Invitrogen™
MA553530 |
100 μL |
£485.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30088457
|
Invitrogen™
MA553531 |
20 μL |
£179.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PEPD Recombinant Monoclonal antibody specifically detects PEPD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| PEPD | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| P12955 | |
| 5184 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2830 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| aminoacyl-L-proline hydrolase; cb1000; dal; fj78g11; Imidodipeptidase; MGC10905; Pep4; Pep-4; Pepd; Peptidase 4; peptidase D; PRD; Prolidase; proline dipeptidase; testicular tissue protein Li 138; wu:fj78g11; xaa-Pro dipeptidase; X-Pro dipeptidase | |
| A synthesized peptide derived from human PRD (461-493AA). | |
| Primary | |
| -20°C | |
| PEPD |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title