missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PEPD Recombinant Rabbit Monoclonal Antibody (23GB2830), Invitrogen™
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA553531
This item is not returnable.
View return policy
Description
Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids. Xaa-Pro dipeptidase is a cytosolic dipeptidase that hydrolyzes dipeptides with proline or hydroxyproline at the carboxy terminus (but not Pro-Pro). It is important in collagen metabolism because of the high levels of iminoacids.
Specifications
| PEPD | |
| Recombinant Monoclonal | |
| 0.8 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P12955 | |
| PEPD | |
| A synthesized peptide derived from human PRD (461-493AA). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2830 | |
| Unconjugated | |
| PEPD | |
| aminoacyl-L-proline hydrolase; cb1000; dal; fj78g11; Imidodipeptidase; MGC10905; Pep4; Pep-4; Pepd; Peptidase 4; peptidase D; PRD; Prolidase; proline dipeptidase; testicular tissue protein Li 138; wu:fj78g11; xaa-Pro dipeptidase; X-Pro dipeptidase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5184 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction