missing translation for 'onlineSavingsMsg'
Learn More

Pentylenetetrazole, TRC

Product Code. 30138208
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30138208 5 g 5g
30134435 10 g 10g
30004239 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30138208 Supplier Toronto Research Chemicals Supplier No. TRCP2275005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-95-5
Molecular Formula C6 H10 N4
Molecular Weight (g/mol) 138.17
Synonym Pentetrazol, 5H-Tetrazolo[1,5-a]azepine, 6,7,8,9-tetrahydro- (8CI, 9CI, ACI), 6,7,8,9-Tetrahydro-5H-tetrazolo[1,5-a]azepine (ACI), 1,2,3,3a-Tetrazacyclohepta-8a,2-cyclopentadiene, 1,5-Pentamethylenetetrazole, 6,7,8,9-Tetrahydro-5H-tetrazoloazepine, 7,8,9,10-Tetrazabicyclo[5.3.0]-8,10-decadiene, Angiazol, Cardiazol, Cardiazole, Cardiol, Cenalene M, Cenazol, Cerebro-nicin, Coranormal, Coranormol, Corasol, Corazol, Corazole, Corazole (analeptic), Corisan, Corsedrol, Cortis, Corvasol, Delzol W, Deumacard, Gewazol, Korazol, Korazole, Leptazol, Leptazole, MeSH ID: D010433, Metrazol, Metrazole, NSC 5729, NSC 66489, Pentamethylenetetrazole, Pentavet, Pentazol, Pentetrazol, Pentetrazole, Pentrazol, Pentrolone, Pentrozol, Pentylenetetrazol, Pentylenetetrazole, Phrenazol, PTZ, Tetracor, Tetrazol, Tetrazole, (1,5-pentanediyl)-, Ventrazol, Yetrazol, α,β-Cyclopentamethylenetetrazole
IUPAC Name 6,7,8,9-tetrahydro-5H-tetrazolo[1,5-a]azepine
SMILES C1CCc2nnnn2CC1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H10N4/c1-2-4-6-7-8-9-10(6)5-3-1/h1-5H2
Formula Weight 138.0905
Percent Purity >95%
Chemical Name or Material Pentetrazol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.