missing translation for 'onlineSavingsMsg'
Learn More

Pentamethylcyclopentadienyltitanium trichloride

Product Code. 11354517
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11354517 Supplier Thermo Scientific Alfa Aesar Supplier No. 039596.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12129-06-5

It is used as a catalyst in Transesterification reactions and styrene polymerization. It is also used as Piano-stool catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12129-06-5
MDL Number MFCD00070434

Specifications

Melting Point 200°C (decomposition)
Color Red
Quantity 5 g
UN Number UN3261
Sensitivity Moisture sensitive
Solubility Information Reacts with water
Formula Weight 289.47
Physical Form Crystalline
Chemical Name or Material Pentamethylcyclopentadienyltitanium trichloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.