accessibility menu, dialog, popup

UserName

H3CCTiClClClCH3CH3CH3H3C

CAS RN 12129-06-5

IUPAC Name:
Synonyms:
Molecular Weight (g/mol): 289.45
Molecular Formula: C10H15Cl3Ti
InChi Key: IPRROFRGPQGDOX-UHFFFAOYSA-K
SMILES: Cl[Ti](Cl)Cl.Cc1c(C)c(C)[c-](C)c1C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results