missing translation for 'onlineSavingsMsg'
Learn More

Pentakis(diethylamino)niobium(V), 99.9% (metals basis), mixture of complexes, Thermo Scientific™

Product Code. 11398398
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398398 500 mg 500mg
11308408 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398398 Supplier Thermo Scientific Alfa Aesar Supplier No. 040445.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Nitride film precursor

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25169-05-5
Molecular Formula C20H50N5Nb
Molecular Weight (g/mol) 453.56
MDL Number MFCD00798536
InChI Key LREVKQSBUDTOJT-UHFFFAOYSA-N
Synonym pentakis diethylamino niobium v
PubChem CID 13981844
IUPAC Name diethylazanide;niobium(5+)
SMILES CCN(CC)[Nb](N(CC)CC)(N(CC)CC)(N(CC)CC)N(CC)CC

Specifications

Boiling Point 120°C (0.1 mmHg)
Odor Amine-like
Linear Formula Nb[N(C2H5)2]5
Quantity 500 mg
UN Number UN3267
Sensitivity Moisture sensitive
Formula Weight 453.56
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Pentakis(diethylamino)niobium(V), Mixture of complexes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.