accessibility menu, dialog, popup

UserName

CAS RN 25169-05-5

NNNNNNbCH3CH3CH3CH3CH3CH3H3CH3CH3CH3C

CAS RN 25169-05-5

IUPAC Name: niobium(5+) pentakis(diethylazanide)
Synonyms: niobium(5+) pentakis(diethylazanide)
Molecular Weight (g/mol): 453.56
Molecular Formula: C20H50N5Nb
InChi Key: LREVKQSBUDTOJT-UHFFFAOYSA-N
SMILES: [Nb+5].CC[N-]CC.CC[N-]CC.CC[N-]CC.CC[N-]CC.CC[N-]CC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)