missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pentafluoropropionaldehyde hydrate, tech. 90%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 50°C to 54°C |
| Linear Formula | CF3CF2CH(OH)2·xH2O |
| Refractive Index | 1.325 |
| Quantity | 5 g |
| Beilstein | 1705233 |
| Formula Weight | 166.05 (Anhydrous) |
| Percent Purity | 90% |
| Grade | Technical |
| Chemical Name or Material | Pentafluoropropionaldehyde hydrate |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering