accessibility menu, dialog, popup

UserName

CAS RN 422-63-9

OFFFFF

CAS RN 422-63-9

IUPAC Name: 2,2,3,3,3-pentafluoropropane-1,1-diol
Synonyms: 2,2,3,3,3-pentafluoropropane-1,1-diol
Molecular Weight (g/mol): 166.05
Molecular Formula: C3H3F5O2
InChi Key: BQAOCAJBGDDVSL-UHFFFAOYSA-N
SMILES: OC(O)C(F)(F)C(F)(F)F
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results