missing translation for 'onlineSavingsMsg'
Learn More

Pentafluoropropionaldehyde hydrate, tech. 90%

Product Code. 11310244
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
11310244 5 g 5g
11320244 25 g 25g
2 options
This item is not returnable. View return policy

Product Code. 11310244

Brand: Thermo Scientific Alfa Aesar L00810.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 422-63-9 | C3HF5O | 148.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 422-63-9
Molecular Formula C3HF5O
Molecular Weight (g/mol) 148.03
MDL Number MFCD00039730
InChI Key IRPGOXJVTQTAAN-UHFFFAOYSA-N
Synonym Pentafluoropropanal hydrate; 2,2,3,3,3-Pentafluoro-1,1-propanediol
IUPAC Name 2,2,3,3,3-pentafluoropropanal
SMILES FC(F)(F)C(F)(F)C=O

Specifications

Melting Point 50°C to 54°C
Linear Formula CF3CF2CH(OH)2·xH2O
Refractive Index 1.325
Quantity 5 g
Beilstein 1705233
Formula Weight 166.05 (Anhydrous)
Percent Purity 90%
Grade Technical
Chemical Name or Material Pentafluoropropionaldehyde hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.